2,2,2-Trifluoro-1-(2-fluoro-3-methoxyphenyl)ethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
64082-500MG
64082-1G
77340-414EA
1033.09
USD
77340-414
77356-460
2,2,2-Trifluoro-1-(2-fluoro-3-methoxyphenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(2-fluoro-3-methoxyphenyl)ethanone
Precaución: for Research Use Only
Formula:
C₉H₆F₄O₂ MW: 222.1363 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24308603 CAS Number: 1500191-80-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...