2,2,2-Trifluoro-1-(2-fluoro-5-methoxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

64081-500MG 64081-1G 64081-5G 64081-25G
77332-074EA 263.68 USD
77332-074 77356-458 77368-160 77389-486
2,2,2-Trifluoro-1-(2-fluoro-5-methoxyphenyl)ethanone ≥97%
2,2,2-Trifluoro-1-(2-fluoro-5-methoxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₆F₄O₂
MW: 222.1363 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26680893
CAS Number: 1505760-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR