2,2,2-Trifluoro-1-(3-(methylthio)phenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

64071-500MG 64071-1G 64071-5G 64071-25G
77332-072EA 85.49 USD
77332-072 77342-232 77368-158 77380-412
2,2,2-Trifluoro-1-(3-(methylthio)phenyl)ethanone ≥97%
2,2,2-Trifluoro-1-(3-(methylthio)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₇F₃OS
MW: 220.2114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11113546
CAS Number: 166831-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR