2,2,2-Trifluoro-1-(3-hydroxy-4-methoxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

64020-500MG 64020-1G 64020-5G
77338-806EA 519.12 USD
77338-806 77356-452 77380-406
2,2,2-Trifluoro-1-(3-hydroxy-4-methoxyphenyl)ethanone ≥97%
2,2,2-Trifluoro-1-(3-hydroxy-4-methoxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₇F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD34761643
CAS Number: 1551034-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR