2,2,2-Trifluoro-1-(4-hydroxy-3-methoxyphenyl)ethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
64014-500MG
64014-1G
64014-5G
77335-882EA
367.71
USD
77335-882
77356-450
77380-404
2,2,2-Trifluoro-1-(4-hydroxy-3-methoxyphenyl)ethanone ≥97%
2,2,2-Trifluoro-1-(4-hydroxy-3-methoxyphenyl)ethanone
Precaución: for Research Use Only
Formula:
C₉H₇F₃O₃ Storage Temperature: Ambient |
MDL Number:
MFCD27665747 CAS Number: 188194-66-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...