1-Ethoxy-2,3-difluoro-5-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

64011-500MG 64011-1G 64011-5G 64011-25G
77332-066EA 225.57 USD
77332-066 77356-446 77368-150 77389-480
1-Ethoxy-2,3-difluoro-5-iodobenzene ≥97%
1-Ethoxy-2,3-difluoro-5-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₇F₂IO
MW: 284.0418 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761638
CAS Number: 2091144-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR