1-(3-Ethoxy-4,5-difluorophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

64007-500MG 64007-1G 64007-5G 64007-25G
77332-060EA 267.8 USD
77332-060 77356-440 77368-144 77389-474
1-(3-Ethoxy-4,5-difluorophenyl)ethanone ≥95%
1-(3-Ethoxy-4,5-difluorophenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O₂
MW: 200.1820 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18375402
CAS Number: 1379306-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR