Ethyl 3-ethoxy-4,5-difluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

64004-500MG 64004-1G 64004-5G
77332-058EA 328.57 USD
77332-058 77356-438 77380-402
Ethyl 3-ethoxy-4,5-difluorobenzoate ≥97%
Ethyl 3-ethoxy-4,5-difluorobenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD34761636
CAS Number: 2385313-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR