Methyl 3-ethoxy-4,5-difluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

64003-500MG 64003-1G 64003-5G
77332-056EA 327.54 USD
77332-056 77356-436 77380-400
Methyl 3-ethoxy-4,5-difluorobenzoate ≥97%
Methyl 3-ethoxy-4,5-difluorobenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD34761635
CAS Number: 2386643-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR