2-Butoxy-3-fluoro-6-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

63990-500MG 63990-1G 63990-5G 63990-25G
77332-052EA 184.37 USD
77332-052 77347-262 77368-140 77389-470
2-Butoxy-3-fluoro-6-methylbenzoic acid ≥95%
2-Butoxy-3-fluoro-6-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₅FO₃
MW: 226.2441 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761625
CAS Number: 2807447-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR