3-Fluoro-2-isopropoxy-6-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

63984-500MG 63984-1G 63984-5G 63984-25G
77332-048EA 283.25 USD
77332-048 77356-428 77368-136 77389-466
3-Fluoro-2-isopropoxy-6-methylbenzaldehyde ≥97%
3-Fluoro-2-isopropoxy-6-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD34761619
CAS Number: 2901096-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR