Methyl 3-fluoro-2-isopropoxy-6-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

63983-500MG 63983-1G 63983-5G 63983-25G
77332-046EA 270.89 USD
77332-046 77356-426 77368-134 77389-464
Methyl 3-fluoro-2-isopropoxy-6-methylbenzoate ≥95%
Methyl 3-fluoro-2-isopropoxy-6-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₅FO₃
MW: 226.2441 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761618
CAS Number: 2901105-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR