2-Ethoxy-1-fluoro-3-iodo-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

63956-500MG 63956-1G 63956-5G
77337-250EA 409.94 USD
77337-250 77356-420 77380-396
2-Ethoxy-1-fluoro-3-iodo-4-methylbenzene ≥97%
2-Ethoxy-1-fluoro-3-iodo-4-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀FIO
MW: 280.0779 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761591
CAS Number: 2385258-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR