4-(Difluoromethoxy)-2,6-dimethylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

63943-500MG 63943-1G 63943-5G
77336-534EA 388.31 USD
77336-534 77356-412 77380-392
4-(Difluoromethoxy)-2,6-dimethylbenzoic acid ≥95%
4-(Difluoromethoxy)-2,6-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O₃
MW: 216.1814 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761579
CAS Number: 2092769-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR