1-(2,5-Difluoro-4-methoxyphenyl)-2,2,2-trifluoroethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
63939-500MG
63939-1G
63939-25G
63939-5G
77332-034EA
301.79
USD
77332-034
77356-410
77389-454
77380-390
1-(2,5-Difluoro-4-methoxyphenyl)-2,2,2-trifluoroethanone ≥97%
1-(2,5-Difluoro-4-methoxyphenyl)-2,2,2-trifluoroethanone
Precaución: for Research Use Only
Formula:
C₉H₅F₅O₂ MW: 240.1268 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD26596071 CAS Number: 1507171-19-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...