2,6-Dimethoxy-4-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

63920-500MG 63920-1G 63920-5G 63920-25G
77332-032EA 225.57 USD
77332-032 77356-408 77368-124 77389-452
2,6-Dimethoxy-4-(methylthio)benzaldehyde ≥97%
2,6-Dimethoxy-4-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃S
Storage Temperature: Ambient
MDL Number: MFCD09030451
CAS Number: 849919-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR