1-(2,5-Dichloro-4-methoxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

63894-500MG 63894-1G 63894-5G 63894-25G
77332-030EA 161.71 USD
77332-030 77345-920 77368-122 77389-450
1-(2,5-Dichloro-4-methoxyphenyl)ethanone ≥97%
1-(2,5-Dichloro-4-methoxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₈Cl₂O₂
MW: 219.0646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21886017
CAS Number: 73239-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR