4-(tert-Butyl)-2-chlorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

63880-500MG 63880-1G 63880-5G 63880-25G
77332-026EA 150.38 USD
77332-026 77345-398 77368-118 77389-446
4-(tert-Butyl)-2-chlorobenzaldehyde ≥97%
4-(tert-Butyl)-2-chlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO
MW: 196.6733 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24686171
CAS Number: 65770-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR