1-(2,6-Difluoro-3-nitrophenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

63867-500MG 63867-1G 63867-5G 63867-25G
77332-022EA 185.4 USD
77332-022 77347-368 77368-116 77389-442
1-(2,6-Difluoro-3-nitrophenyl)ethanone ≥97%
1-(2,6-Difluoro-3-nitrophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₅F₂NO₃
MW: 201.1270 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23844884
CAS Number: 91188-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR