2,2,2-trifluoro-1-(2-methoxy-6-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

63858-500MG 63858-1G 63858-5G 63858-25G
77332-018EA 215.27 USD
77332-018 77356-400 77368-112 77389-440
2,2,2-trifluoro-1-(2-methoxy-6-methylphenyl)ethanone ≥95%
2,2,2-trifluoro-1-(2-methoxy-6-methylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₂
MW: 218.1724 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761555
CAS Number: 1547136-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR