2,2,2-Trifluoro-1-(5-methoxy-2-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

63852-500MG 63852-1G 63852-5G 63852-25G
77332-016EA 258.53 USD
77332-016 77356-396 77368-110 77389-438
2,2,2-Trifluoro-1-(5-methoxy-2-methylphenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(5-methoxy-2-methylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₂
MW: 218.1724 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31697048
CAS Number: 2293688-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR