2,2,2-Trifluoro-1-(3-(methoxymethyl)phenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

63847-500MG 63847-1G 63847-5G 63847-25G
77332-012EA 241.02 USD
77332-012 77356-392 77368-106 77389-434
2,2,2-Trifluoro-1-(3-(methoxymethyl)phenyl)ethanone ≥97%
2,2,2-Trifluoro-1-(3-(methoxymethyl)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₂
MW: 218.1724 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31621287
CAS Number: 1610418-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR