3-Methyl-4-(2,2,2-trifluoroacetyl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

63839-500MG 63839-1G 63839-5G 63839-25G
77332-010EA 190.55 USD
77332-010 77356-388 77368-104 77389-432
3-Methyl-4-(2,2,2-trifluoroacetyl)benzonitrile ≥97%
3-Methyl-4-(2,2,2-trifluoroacetyl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₀H₆F₃NO
MW: 213.1559 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28502950
CAS Number: 2089726-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR