2,2,2-Trifluoro-1-(2-fluoro-3-hydroxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

63829-500MG 63829-1G 63829-5G 63829-25G
77332-006EA 202.91 USD
77332-006 77356-386 77368-100 77389-430
2,2,2-Trifluoro-1-(2-fluoro-3-hydroxyphenyl)ethanone ≥97%
2,2,2-Trifluoro-1-(2-fluoro-3-hydroxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₄F₄O₂
MW: 208.1097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26687890
CAS Number: 1508887-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR