4-Bromo-3-fluoro-2-(methylthio)phenol ≥97%

Supplier: AOBChem USA

Warning

63804-500MG 63804-1G 63804-5G
77331-996EA 319.3 USD
77331-996 77356-370 77380-362
4-Bromo-3-fluoro-2-(methylthio)phenol ≥97%
4-Bromo-3-fluoro-2-(methylthio)phenol

Precaución: for Research Use Only

Formula: C₇H₆BrFOS
Storage Temperature: Ambient
MDL Number: MFCD28739505
CAS Number: 1807118-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR