1-(5-(Ttert-butyl)-2-ethoxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

63795-500MG 63795-1G 63795-5G 63795-25G
77331-992EA 257.5 USD
77331-992 77356-368 77368-092 77389-424
1-(5-(Ttert-butyl)-2-ethoxyphenyl)ethanone ≥97%
1-(5-(Ttert-butyl)-2-ethoxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₂
MW: 220.3074 g/mol
Storage Temperature: Ambient
CAS Number: 932831-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR