(3-Ethoxy-5-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

63779-500MG 63779-1G 63779-5G
77340-214EA 836.36 USD
77340-214 77356-364 77380-360
(3-Ethoxy-5-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%
(3-Ethoxy-5-(trifluoromethyl)phenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₁F₃OS
MW: 236.2539 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761524
CAS Number: 2807450-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR