3-(5-Fluoro-2-methoxyphenyl)-3-hydroxypropanoic acid ≥95%

Supplier: AOBChem USA

Warning

63774-500MG 63774-1G 63774-5G
77339-618EA 603.58 USD
77339-618 77356-360 77380-356
3-(5-Fluoro-2-methoxyphenyl)-3-hydroxypropanoic acid ≥95%
3-(5-Fluoro-2-methoxyphenyl)-3-hydroxypropanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₄
MW: 214.1903 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761519
CAS Number: 1499301-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR