3-(4-Bromo-3-(trifluoromethyl)phenyl)-3-hydroxypropanoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
63773-500MG
63773-1G
63773-5G
77339-488EA
593.28
USD
77339-488
77356-358
77380-354
3-(4-Bromo-3-(trifluoromethyl)phenyl)-3-hydroxypropanoic acid ≥97%
3-(4-Bromo-3-(trifluoromethyl)phenyl)-3-hydroxypropanoic acid
Precaución: for Research Use Only
Formula:
C₁₀H₈BrF₃O₃ MW: 313.0679 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34761518 CAS Number: 2353408-10-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...