3-(4-Bromo-2-(trifluoromethyl)phenyl)-3-hydroxypropanoic acid ≥95%

Supplier: AOBChem USA

Warning

63772-500MG 63772-1G 63772-5G
77338-756EA 516.03 USD
77338-756 77356-356 77380-352
3-(4-Bromo-2-(trifluoromethyl)phenyl)-3-hydroxypropanoic acid ≥95%
3-(4-Bromo-2-(trifluoromethyl)phenyl)-3-hydroxypropanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₈BrF₃O₃
MW: 313.0679 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761517
CAS Number: 2353541-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR