3-(2-Bromo-6-chlorophenyl)-3-hydroxypropanoic acid ≥95%

Supplier: AOBChem USA

Warning

63771-500MG 63771-1G 63771-5G
77339-616EA 603.58 USD
77339-616 77356-354 77380-350
3-(2-Bromo-6-chlorophenyl)-3-hydroxypropanoic acid ≥95%
3-(2-Bromo-6-chlorophenyl)-3-hydroxypropanoic acid

Precaución: for Research Use Only

Formula: C₉H₈BrClO₃
MW: 279.5150 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761516
CAS Number: 2228159-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR