3-(4-Bromo-2-ethylphenyl)-3-hydroxypropanoic acid ≥97%

Supplier: AOBChem USA

Warning

63768-500MG 63768-1G
77340-510EA 1364.75 USD
77340-510 77356-350
3-(4-Bromo-2-ethylphenyl)-3-hydroxypropanoic acid ≥97%
3-(4-Bromo-2-ethylphenyl)-3-hydroxypropanoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO₃
MW: 273.1231 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761514
CAS Number: 2807464-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR