2,2,2-Trifluoro-1-(2-hydroxy-3-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

63765-500MG 63765-1G
77340-436EA 1056.78 USD
77340-436 77356-348
2,2,2-Trifluoro-1-(2-hydroxy-3-methylphenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(2-hydroxy-3-methylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₇F₃O₂
MW: 204.1458 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761512
CAS Number: 2353534-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR