2,2,2-Trifluoro-1-(4-hydroxy-3-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

63759-500MG 63759-1G 63759-5G
77340-166EA 806.49 USD
77340-166 77356-344 77380-344
2,2,2-Trifluoro-1-(4-hydroxy-3-methylphenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(4-hydroxy-3-methylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₇F₃O₂
MW: 204.1458 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34169042
CAS Number: 1554485-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR