1-(3-Ethylphenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

63755-500MG 63755-1G 63755-5G 63755-25G
77331-986EA 155.53 USD
77331-986 77345-576 77368-088 77389-420
1-(3-Ethylphenyl)-2,2,2-trifluoroethanone ≥97%
1-(3-Ethylphenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O
Storage Temperature: Ambient
MDL Number: MFCD11113543
CAS Number: 155628-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR