1-(2,5-Dimethylphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

63752-500MG 63752-1G 63752-5G 63752-25G
77331-982EA 92.7 USD
77331-982 77342-470 77368-084 77380-340
1-(2,5-Dimethylphenyl)-2,2,2-trifluoroethanone ≥95%
1-(2,5-Dimethylphenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O
MW: 202.1730 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01319985
CAS Number: 181828-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR