1-(Benzyloxy)-2-chloro-3-fluoro-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

63742-500MG 63742-1G 63742-5G
77338-576EA 499.55 USD
77338-576 77356-340 77380-336
1-(Benzyloxy)-2-chloro-3-fluoro-5-methylbenzene ≥97%
1-(Benzyloxy)-2-chloro-3-fluoro-5-methylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂ClFO
MW: 250.6958 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761506
CAS Number: 2807464-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR