1-Chloro-4-fluoro-5-iodo-2-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

63740-500MG 63740-1G 63740-5G 63740-25G
77331-980EA 227.63 USD
77331-980 77356-338 77368-082 77389-418
1-Chloro-4-fluoro-5-iodo-2-methoxybenzene ≥97%
1-Chloro-4-fluoro-5-iodo-2-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅ClFIO
Storage Temperature: Ambient
MDL Number: MFCD34761505
CAS Number: 2383863-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR