(5-Chloro-2-fluoro-4-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

63739-500MG 63739-1G 63739-5G 63739-25G
77331-978EA 224.54 USD
77331-978 77356-336 77368-080 77389-416
(5-Chloro-2-fluoro-4-methoxyphenyl)(methyl)sulfane ≥97%
(5-Chloro-2-fluoro-4-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈ClFOS
MW: 206.6649 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761504
CAS Number: 2710759-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR