1-(5-Chloro-2-fluoro-4-methoxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

63735-500MG 63735-1G 63735-5G 63735-25G
77331-976EA 206 USD
77331-976 77356-334 77368-078 77389-414
1-(5-Chloro-2-fluoro-4-methoxyphenyl)ethanone ≥97%
1-(5-Chloro-2-fluoro-4-methoxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₈ClFO₂
MW: 202.6100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28359921
CAS Number: 1783421-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR