(5-Chloro-4-ethoxy-2-fluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

63733-500MG 63733-1G 63733-5G
77338-574EA 499.55 USD
77338-574 77356-330 77380-332
(5-Chloro-4-ethoxy-2-fluorophenyl)(methyl)sulfane ≥97%
(5-Chloro-4-ethoxy-2-fluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₀ClFOS
MW: 220.6915 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761502
CAS Number: 2706964-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR