5-Chloro-4-ethoxy-2-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

63727-500MG 63727-1G
77340-470EA 1199.95 USD
77340-470 77356-328
5-Chloro-4-ethoxy-2-fluorobenzaldehyde ≥97%
5-Chloro-4-ethoxy-2-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈ClFO₂
MW: 202.6100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06246183
CAS Number: 883521-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR