1-(Benzyloxy)-2-(tert-butyl)-4-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

63723-500MG 63723-1G 63723-5G 63723-25G
77331-974EA 168.92 USD
77331-974 77346-332 77368-076 77389-412
1-(Benzyloxy)-2-(tert-butyl)-4-iodobenzene ≥95%
1-(Benzyloxy)-2-(tert-butyl)-4-iodobenzene

Precaución: for Research Use Only

Formula: C₁₇H₁₉IO
MW: 366.2366 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761501
CAS Number: 330935-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR