(4-(Benzyloxy)-3-(tert-butyl)phenyl)(methyl)sulfane ≥97%
Supplier: AOBChem USA
|
Warning
|
63722-500MG
63722-1G
63722-5G
63722-25G
77331-972EA
186.43
USD
77331-972
77356-324
77368-074
77389-410
(4-(Benzyloxy)-3-(tert-butyl)phenyl)(methyl)sulfane ≥97%
(4-(Benzyloxy)-3-(tert-butyl)phenyl)(methyl)sulfane
Precaución: for Research Use Only
Formula:
C₁₈H₂₂OS Storage Temperature: Ambient |
MDL Number:
MFCD34761500 CAS Number: 2807455-25-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...