4-(Benzyloxy)-3-(tert-butyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

63713-500MG 63713-1G 63713-5G
77338-320EA 476.89 USD
77338-320 77356-322 77380-330
4-(Benzyloxy)-3-(tert-butyl)benzoic acid ≥97%
4-(Benzyloxy)-3-(tert-butyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₈H₂₀O₃
MW: 284.3496 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19439973
CAS Number: 146852-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR