1-Ethoxy-2,4-difluoro-5-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

63699-500MG 63699-1G 63699-5G 63699-25G
77331-968EA 155.53 USD
77331-968 77345-574 77368-068 77389-406
1-Ethoxy-2,4-difluoro-5-methoxybenzene ≥97%
1-Ethoxy-2,4-difluoro-5-methoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀F₂O₂
MW: 188.1713 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761491
CAS Number: 2386670-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR