1,5-Difluoro-2,4-dimethoxybenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
1,5-Difluoro-2,4-dimethoxybenzene, 4,6-Difluororesorcinol dimethyl ether
63698-500MG
63698-1G
63698-5G
63698-25G
77331-966EA
91.67
USD
77331-966
77342-436
77368-066
77380-324
1,5-Difluoro-2,4-dimethoxybenzene ≥97%
1,5-Difluoro-2,4-dimethoxybenzene
Precaución: for Research Use Only
Formula:
C₈H₈F₂O₂ MW: 174.145 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13193047 CAS Number: 79069-70-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...