(2,4-Difluoro-3-iodo-5-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

63697-500MG 63697-1G 63697-5G
77338-160EA 466.59 USD
77338-160 77356-318 77380-322
(2,4-Difluoro-3-iodo-5-methoxyphenyl)(methyl)sulfane ≥97%
(2,4-Difluoro-3-iodo-5-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₇F₂IOS
MW: 316.1068 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761490
CAS Number: 2807470-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR