(2,6-Difluoro-3-methoxy-5-(methylthio)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

63696-500MG 63696-1G 63696-5G
77339-088EA 548.99 USD
77339-088 77356-316 77380-320
(2,6-Difluoro-3-methoxy-5-(methylthio)phenyl)methanol ≥97%
(2,6-Difluoro-3-methoxy-5-(methylthio)phenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₀F₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD34761489
CAS Number: 2807447-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR