2,6-Difluoro-3-methoxy-5-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

63693-500MG 63693-1G 63693-5G
77336-262EA 384.19 USD
77336-262 77356-314 77380-318
2,6-Difluoro-3-methoxy-5-(methylthio)benzoic acid ≥97%
2,6-Difluoro-3-methoxy-5-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₈F₂O₃S
Storage Temperature: Ambient
MDL Number: MFCD34761486
CAS Number: 2807471-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR